missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilic Acid, TRC

Product Code. 17943375
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17943375 5 g 5g
17971555 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17943375 Supplier Toronto Research Chemicals Supplier No. TRCS6890955G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-57-3
Molecular Formula C6 H7 N O3 S
Molecular Weight (g/mol) 173.19
Synonym 4-Aminobenzenesulfonic Acid, Sulfanilic Acid, Sulphanilic Acid, Sulfadimidine Imp. F (Pharmeuropa), Sulfadiazine Imp. B (Pharmeuropa), Sulfamethoxazole Imp. D (EP), Mesalazine Imp. O (EP)
IUPAC Name 4-aminobenzenesulfonic acid
SMILES Nc1ccc(cc1)S(=O)(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H7NO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H,8,9,10)
Formula Weight 173.0147
Chemical Name or Material Sulfanilic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.