missing translation for 'onlineSavingsMsg'
Learn More

Sulfasalazine, Dr. Ehrenstorfer

Product Code. 30311237
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311237 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311237 Supplier Dr. Ehrenstorfer Supplier No. DREC17000150
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfasalazine
CAS 599-79-1
Analyte or Component Names Sulfasalazine
Shipping Condition +20°C
Molecular Formula C18 H14 N4 O5 S
Certifications/Compliance ISO/IEC 17025
Synonym Sulfasalazine, 2-Hydroxy-5-[2-[4-(pyridin-2-ylsulfamoyl)phenyl]diazenyl]benzoic acid, Benzoic acid, 2-hydroxy-5-[2-[4-[(2-pyridinylamino)sulfonyl]phenyl]diazenyl]-Benzoic acid, 2-hydroxy-5-[[4-[(2-pyridinylamino)sulfonyl]phenyl]azo]- (9CI), Salicylic acid, 5-[[p-(2-pyridylsulfamoyl)phenyl]azo]- (6CI,7CI,8CI), 2-Hydroxy-5-((4-((2-pyridinylamino)sulfonyl)phenyl)azo)benzoic acid, 5-[4-(2-Pyridylsulfamyl)phenylazo]-2-hydroxybenzoic acid, 5-[p-(2-Pyridylsulfamoyl)phenylazo]salicylic acid, 5-[p-(2-Pyridylsulfamyl)phenylazo]salicylic acid, Azopyrin, Azopyrine, Azulfidine, Azulfidine EN, Azulfin, Benzosulfa, Colo-Pleon, NSC 203730, NSC 667219, Reupirin, Salazopyridin, Salazopyrin, Salazopyrine, Salazosulfapyridin, Salazosulfapyridine, Salicylazosulfapyridine, Salisulf, Sulfasalazin, Sulfasalazine, Sulphasalazine
InChI Formula InChI=1S/C18H14N4O5S/c23-16-9-6-13(11-15(16)18(24)25)21-20-12-4-7-14(8-5-12)28(26,27)22-17-3-1-2-10-19-17/h1-11,23H,(H,19,22)(H,24,25)/b21-20+
SMILES OC(=O)c1cc(ccc1O)N=Nc2ccc(cc2)S(=O)(=O)Nc3ccccn3
IUPAC Name 2-hydroxy-5-[(E)-[4-(pyridin-2-ylsulfamoyl)phenyl]diazenyl]benzoic acid
Molecular Weight (g/mol) 398.39
Quantity 100 mg
Formula Weight 398.0685 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.