missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Certified Reference Material, UKAS ISO 17025 & ISO 17034 Certified, Sulfur in heavy mineral oil, 0.5000 % (5000ppm), 100mL., Paragon Scientific™

Product Code. 16020252
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16020252 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16020252 Supplier Paragon Scientific Supplier No. HMSU14
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Suitable for calibration, verification or for use in quality control procedures used for analysing sulfur content in petroleum products, with sulfur content ranging from 0 (Blank) through to 5.0000% (50000ppm) for each material.

  • Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation.
  • Highest level of accreditation guarantee, providing the most credible certified data available worldwide.
  • Suitable for but not limited to internationally recognised test methods and protocol: ASTM D2622, ASTM D4294, ASTM D5453, ASTM D7039, IP 336, IP 496, IP 497.
  • Fully traceable to international standards and low levels of uncertainty achieves maximum accuracy of data.
  • Supplied in 100 mL volume, tamper-evident glass packaging, providing assurance of sample integrity.
  • Manufactured in the United Kingdom.
  • 12 month shelf life.

Specifications

Quantity 100 mL

Nominal values quoted. Certified Values will vary from batch to batch.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.