missing translation for 'onlineSavingsMsg'
Learn More

Sulfur and Nitrogen Standard: S @ 0, 1, 5, 10, 15, 20 ng/μL and N @ 0, 1, 5, 20, 35, 50 ng/μL in Isooctane

Product Code. 30326153
Click to view available options
Quantity:
Set (6)
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30326153

Brand: VHG Labs VHGSNSET1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitrogen∣Sulfur
CAS 7727-37-9, 7704-34-9
Analyte or Component Names Nitrogen∣Sulfur
Shipping Condition Room Temperature
Solution Type Mixture
Element Multi-element
Solvent or Matrix Isooctane
Industry Type Petrochemical Testing
Quantity Set (6)
Concentration or Composition (by Analyte or Components) Sulfur 0, 1, 5, 10, 15, 20 ng/μL∣Nitrogen 0, 1, 5, 20, 35, 50 ng/μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.