missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur and Nitrogen Standard: S @ 0, 1, 5, 10, 15, 20 ng/μL and N @ 0, 1, 5, 20, 35, 50 ng/μL in Isooctane, VHG™
Click to view available options
Quantity:
6 x 2 mL
Unit Size:
12mL
Description
Sulfur and Nitrogen Standard: S @ 0, 1, 5, 10, 15, 20 ng/μL and N @ 0, 1, 5, 20, 35, 50 ng/μL in Isooctane
Specifications
Specifications
| Chemical Name or Material | Nitrogen/Sulfur |
| CAS | 7727-37-9, 7704-34-9 |
| Analyte or Component Names | Nitrogen/Sulfur |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Isooctane |
| Quantity | 6 x 2 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction