missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 1M, Chem-Lab

Product Code. 11963004
Change view
Click to view available options
Quantity:
1 L
Packaging:
Polyethylene
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11963004 1 L Polyethylene 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11963004 Supplier Chem-Lab Supplier No. CL0526581000
  • ONLINE EXCLUSIVE PRICE
    31.00€
    18.39€ / 1 L
    Save 12.61€
    40% Off
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for titrations and TMAR silicon determinations. Sulfuric Acid, 1M, Chem-Lab is a dilute solution of 98.08g H2SO4 per liter of water for use in volumetric analysis and other laboratory applications.

Sulfuric acid is a highly corrosive, strong mineral acid that shows different properties depending upon its concentration. It is a pungent-ethereal, colorless to slightly yellow viscous liquid. Sulfuric acid is a diprotic acid that is hygroscopic. Concentrated sulfuric acid has strong dehydrating and oxidizing properties. Dilute sulfuric acid is a good electrolyte.

Specifications

pH <2
Color Colorless
CAS 7664-93-9, 7732-18-5
Composition 98.08g H2SO4 per 1L H2O
Melting Point 0°C
Boiling Point 100°C
Concentration 1mol/L
Density 1.08g/mL
Odor Odorless
Packaging Polyethylene
Normality 2N (±0.004, 20°C)
Quantity 1 L
Molecular Formula H2O4S
Viscosity 1.14 cSt
Physical Form Liquid
MDL Number MFCD00064589
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
Solubility Information Soluble in Water
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Formula Weight 98.08g/mol
Show More Show Less
compliance-icons
Product Identifier
  • Sulfuric acid 1 mol/l
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.