Learn More
Buffer Solution, pH 7.00, Chem-Lab
Product Code. 10799994
Click to view available options
Quantity:
1 L
5 L
Packaging:
Ethylene propylene
Polyethylene
Unit Size:
1 L
5 L
Product Code. 10799994
Brand: Chem-Lab CL03.0216.1000
Edge
Designed for pH measurement and maintenance in pH-neutral samples. Buffer Solution, pH 7.00, Chem-Lab is a phosphate solution with a pH level accurate to ±0.02. The solution is traceable to Standard Reference Materials (SRM) from NIST.
pH Levels ±0.02:
  • pH 7.13 — 0°C
  • pH 7.05 — 10°C
  • pH 7.00 — 20°C
  • pH 6.99 — 25°C
  • pH 6.98 — 30°C
  • pH 6.97 — 40°C
  • pH 6.96 — 50°C
  • pH 6.96 — 60°C
  • pH 6.97 — 70°C
  • pH 6.98 — 80°C
  • pH 7.00 — 90°C
Specifications
Melting Point 0°C
Boiling Point 100°C
Color Colorless
Density 1.01g/mL
For Use With (Application) Laboratory use, pH measurements
pH 7.00 ±0.02
Physical Form Liquid
Quantity 1 L
Solution Type pH Buffer
Formulation Phosphate
Odor Odorless
Packaging Polyethylene
Traceability to NIST Yes
Solubility Information Soluble in water