missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulphuric Acid 0.25 M (0.5 N) Analytical Volumetric Solution (AVL), Reagecon
Shop All Reagecon Products

Click to view available options
:
5 L
Description
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- Accurate, stable, reproducible and fully factorized.
- Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
- Stability is proven (either opened or unopened) for the shelf life of this product.
- Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
- Certified to a specification of ± 0.1%.
- Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
- May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
- Acid/base titrations, also called neutralization titrations, are used to determine either the amount of acid/base in a sample or the substance that can be converted to an acid/base.
Specifications
Specifications
| Concentration | 0.25 M (0.5 N) |
| Packaging | HDPE Container |
| Quantity | 5 L |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction