missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sumatriptan Succinate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Sumatriptan Succinate |
| CAS | 103628-48-4 |
| Analyte or Component Names | Sumatriptan Succinate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H21 N3 O2 S . C4 H6 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Sumatriptan Succinate, GR 43175C, Sumatriptan succinate (GR 43175C), Ph Eur Sumatriptan Impurity Mixture, GR 43175X, Sumatriptan Test Mix for Impurity Identification Solution 2 |
| InChI Formula | InChI=1S/C14H21N3O2S.C4H6O4/c1-15-20(18,19)10-11-4-5-14-13(8-11)12(9-16-14)6-7-17(2)3;5-3(6)1-2-4(7)8/h4-5,8-9,15-16H,6-7,10H2,1-3H3;1-2H2,(H,5,6)(H,7,8) |
| Solution Type | Neat |
| SMILES | CNS(=O)(=O)Cc1ccc2[nH]cc(CCN(C)C)c2c1.OC(=O)CCC(=O)O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction