Learn More
SUMO1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 450.00€
Specifications
| Antigen | SUMO1 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18030736
|
Proteintech
CL488-10329-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18038085
|
Proteintech
CL488-10329-100UL |
100 μL |
450.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SUMO1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SUMO1. This antibody recognizes human, mouse, rat antigen. The SUMO1 antibody has been validated for the following applications: FC, IF analysis. This antibody is coralite™ plus 488.
Specifications
| SUMO1 | |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human, Mouse, Rat | |
| DAP 1, GAP modifying protein 1, GMP1, OFC10, PIC1, SENP2, Sentrin, SMT3, SMT3 homolog 3, SMT3C, SMT3H3, SUMO 1, SUMO1, Ubiquitin like protein SMT3C, Ubiquitin like protein UBL1, UBL1 | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 7341 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.