missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
199.00€ - 351.00€
Specifications
| Antigen | SUMO2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18023814
|
Novus Biologicals
NB100-56444SS |
0.025 mg |
199.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18706323
|
Novus Biologicals
NB100-56444 |
0.1 mg |
351.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SUMO2 Polyclonal specifically detects SUMO2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SUMO2 | |
| Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| P61956 | |
| 6613 | |
| The amino acid sequences used as immunogen for this antibody are 4-16 (EKPKEGVKTENDH) of human SUMO2 (corresponding to amino acids 4-16 of SUMO3) and 61-76 (RFDGQPINETDTPAQL) of human SUMO2 (corresponding to amino acids 60-75 of SUMO3). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Epigenetics | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| HSMT3, MGC117191, sentrin 2, Sentrin-2, small ubiquitin-like modifier 2, small ubiquitin-related modifier 2, SMT3 (suppressor of mif two 3, yeast) homolog 2, SMT3 homolog 2, SMT3 suppressor of mif two 3 homolog 2 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 2 (yeast), SMT3A, SMT3B, SMT3H2, SUMO-2, SUMO3, SUMO-3, Ubiquitin-like protein SMT3A, ubiquitin-like protein SMT3B | |
| SUMO2 | |
| IgG | |
| Protein G purified | |
| Based on homology of the amino acids used as immunogen, this antibody will detect human and mouse SUMO2 and human and mouse SUMO3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title