Learn More
Thermo Scientific™ SUN-SRi™ SunFlo™ Dissolution Filters
Manufactured of Ultra High Molecular Weight (UHMW) foamed porous polyethylene
Brand: Thermo Scientific™ 14823272
Additional Details : Weight : 0.80000kg
Description
- Full flow design removes small particulates
- Fit securely on the end of a 1/8 in. cannula or sample probe
Specifications
45 μm | |
Case of 1000 | |
1/8 in. Cannula or Probe |