Learn More
Thermo Scientific™ SUN-SRi™ SunFlo™ Dissolution Filters
Manufactured of Ultra High Molecular Weight (UHMW) foamed porous polyethylene
POA
Specifications
For Use With 1/8 in. Cannula or Probe
View More Specs
Products 1
Product Code Brand Porosity Packaging Quantity Price Quantity & Availability  
Product Code Brand Porosity Packaging Quantity Price Quantity & Availability  
11922005
View Documents Promotion Details
Thermo Scientific™
14823272
45 μm Case of 1000
POA
Pack of 1000
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • Full flow design removes small particulates
  • Fit securely on the end of a 1/8 in. cannula or sample probe
Specifications
Specifications
1/8 in. Cannula or Probe