missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript V Reaction Buffer

Product Code. 32260513
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32260513 5 x 1 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32260513 Supplier Invitrogen™ Supplier No. 19090010B
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Invitrogen™ SuperScript™ V Reaction Buffer is specifically formulated for use with SuperScript™ V Reverse Transcriptase (RT) to support efficient and reliable first-strand cDNA synthesis.

Invitrogen™ SuperScript™ V Reaction Buffer is specifically formulated for use with SuperScript™ V Reverse Transcriptase (RT) to support efficient and reliable first-strand cDNA synthesis. The buffer is supplied with SuperScript V RT and is also available separately for applications requiring additional reaction buffer.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
For Use With (Application) Reverse Transcription
pH Not Available
Quantity 5 x 1 mL
Includes SuperScript V Reaction Buffer (5 x 1 mL)

For Research Use Only. Not for use in diagnostic procedures.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.