Learn More
Thermo Scientific™ SuperSealer
Product Code. 10758640
Change view
Click to view available options
Product Type:
Mat
Microplate Sealer
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
10758640 Microplate Sealer Each
11932161 Mat Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10758640 Supplier Thermo Scientific™ Supplier No. 411011
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ensure a proper seal on plates, blocks and storage tube racks with the Thermo Scientific™ SuperSealer.
Ensure a proper seal on plates, blocks and storage tube racks with the Thermo Scientific™ SuperSealer. Choose individual sealing options such as Thermo Scientific™ Matrix™ SepraSeal mats for liquid handling and single tube accessibility, or seal entire plates and blocks for storage with Thermo Scientific™ Nunc™ CapMats. Effectively apply adhesive tape seals and ensure a tight seal with the included SuperSealer Adhesive Seal Mat.
Sealing Simplicity
  • Simplifies the sealing of plates, blocks and tube racks of virtually any height, with no adjustments needed, ensuring a secure and consistent seal
  • Once the seal along with the plate, block or tube rack are placed into the drawer, simply close the door to initiate sealing
Sealing Flexibility
  • Designed to work with Thermo Scientific™ Matrix™ SepraSeal mats, CapMats and, with the SuperSealer AdhesiveSeal Mat, any pre-cut adhesive tape seal, you can be assured a proper seal regardless of sealing option
Includes: Automatic cap, tape and SepraSeal mat applicator
TRUSTED_SUSTAINABILITY
Specifications
Description Thermo Scientific Super Sealer
No. per Pack 1 Ea.
Product Type Microplate Sealer