Learn More
Thermo Scientific Support Bridge
Product Code. 10527305 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
2 L BioBottle
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10527305 2 L BioBottle Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10527305 Supplier Thermo Scientific Supplier No. 75007686
  • ONLINE EXCLUSIVE PRICE
    253.00€
    234.00€ / Pack of 2
    Save 19.00€
    7% Off
View all promotions
Estimated Shipment: 25-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Support Bridge for 2 L BioBottle.
  • Support bridge (x2) for 2 L BioBottle
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 2 L BioBottle
Disposable Reusable
Quantity 2 Pk.
Product Type Support Bridge