missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUR-8 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99515DL594
This item is not returnable.
View return policy
Description
SUR-8 Polyclonal antibody specifically detects SUR-8 in Human samples. It is validated for Western Blot
Specifications
| SUR-8 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 594 | |
| KIAA0862FLJ60412, Protein soc-2 homolog, Protein sur-8 homolog, SIAA0862, SOC-2, soc-2 (suppressor of clear, C.elegans) homolog, soc-2 suppressor of clear homolog (C. elegans), SOC2SUR8leucine-rich repeat protein SHOC-2, SUR-8 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SUR-8. | |
| 0.1 mL | |
| Signal Transduction | |
| 8036 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction