missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureCast™ Gel Spacer

Product Code. 15503381
Change view
Click to view available options
Quantity:
10 spacers
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15503381 10 spacers Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15503381 Supplier Invitrogen™ Supplier No. HC1003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Leak-free, confident handcasting of your polyacrylamide gels with the Invitrogen™ SureCast™ Handcast System.

The SureCast Gel Spacers are a replacement set of sealing gaskets for use with the SureCast Glass Plates to create a leak-free cassette used in the SureCast Handcast Station. Comes in a package of 10 gel spacers.
  • Compatible only with the SureCast Handcast System

Recommended Storage

Room Temperature

Warranty

Up to 12 months

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) SureCast Glass Plate
Gel Size Mini
Product Line SureCast
Type SureCast™ Gel Spacer
Quantity 10 spacers
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.