Learn More
Tork Surface Disinfectant Spray
Product Code. 30243768 Shop All Tork Products
Change view
Click to view available options
Scent:
Unscented
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Scent Unit Size
30243768 Unscented Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30243768 Supplier Tork Supplier No. 193610
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Tork Surface Disinfecting Spray is a ready to use disinfectant that is effective against a broad spectrum of bacteria, yeast and viruses, including coronavirus.
  • Kills 99.99% of enveloped viruses in five minutes including Coronavirus EN14476: Limited virucidal activity (active against enveloped viruses such as, e.g. Influenza Virus, Coronavirus, HIV, Herpes Virus, Hepatitis Virus) exposure time: 5 minutes under dirty conditions.
  • Biodegradable formula. 100% plant-based active ingredient (lactic acid).
  • Food surface contact safe The Surface Disinfecting Spray is suitable for PT2 (disinfectants not intended for direct application to humans or animals) and PT4 (food and feed area) applications, food contact surfaces must be rinsed with drinking water after disinfecting with product.
  • The product is an alcohol-free ready-to-use multi-purpose spray for surface disinfection.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Trigger Spray Bottle
Color-coded No
Form Clear Liquid
Scent Unscented
Sterility Non-sterile
Color Clear
Packaging Plastic Bottle
Alternate Part Number 7322541432632