Learn More
Tork Surface Disinfectant Wet Wipes
Product Code. 30243842 Shop All Tork Products
Change view
Click to view available options
Scent:
Unscented
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Scent unitSize
30243842 Unscented Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30243842 Supplier Tork Supplier No. 193600
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
Tork Surface Disinfecting Wet Wipes are a ready to use disinfectant that is effective against a broad spectrum of bacteria, yeast and viruses, including coronavirus.
  • Kills 99.99% enveloped viruses in 30 seconds including Coronavirus EN14476: Virucidal activity (Limited virucidal, so all enveloped viruses such as Influenza virus, Coronavirus, HIV, Herpes virus, Hepatitis virus). Exposure time: 30 seconds under clean conditions.
  • Dermatologically tested. The product is dermatologically tested but intended exclusively for use on surfaces and objects.
  • Ethanol free
  • Resealable lid to help keep wipes moist
TRUSTED_SUSTAINABILITY
Specifications
Product Type Wipes
Color-coded No
Form Sachet Wipe
Scent Unscented
Sterility Non-sterile
Color White
Disposable Limited-use
Ply 1-Ply