Learn More
Terumo™ SurGuard2™ Hypodermic Safety Needles
Designed with safety in mind, these hypodermic needles include a simple locking mechanism to reduce needle stick injuries. Terumo™ SurGuard2™ Hypodermic Safety Needles can be easily activated using only one hand.
Brand:  Terumo™ 8SG22516MP
Additional Details : Weight : 0.24300kg
 View more versions of this product
Product Code. 12661146
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Simple locking mechanism for safety
  • Safety lock can be activated using only one hand
  • Standard hub equally fits to all luer lock and luer slip syringes
Specifications
Specifications
25 G
Hypodermic Needle
16 mm
Safety lock
800/Pk.