missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUV420h1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99518-50ul
This item is not returnable.
View return policy
Description
SUV420h1 Polyclonal antibody specifically detects SUV420h1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SUV420h1 | |
| Polyclonal | |
| Immunohistochemistry 1:500-1:2000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| C630029K18Rik, CGI85, CGI-85, EC 2.1.1.43, histone-lysine N-methyltransferase SUV420H1, KMT5B, KMT5B Lysine N-methyltransferase 5B, MGC118906, MGC118909, MGC21161, MGC703, Su(var)4-20 homolog 1, Suppressor of variegation 4-20 homolog 1, suppressor of variegation 4-20 homolog 1 (Drosophila), suv4-20h1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SUV420h1. | |
| 50 μg | |
| Cancer, Epigenetics, Transcription Factors and Regulators | |
| 51111 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering