Learn More
SVL™ Bearing Stirrer for SVL™ Stirrer Shaft
Product Code. 11937298
Change view
Click to view available options
Standard Taper Joint:
19/26
29/32
30
45/40
Unit Size:
Each
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Unit Size
11937298 29/32 Each
15527730 30 Pack of 5
15537730 19/26 Pack of 5
15557730 45/40 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11937298 Supplier SVL™ Supplier No. 58809
  • ONLINE EXCLUSIVE PRICE
    284.55€
    203.96€ / Pack of 1
    Save 80.59€
    28% Off
View all promotions
Estimated Shipment: 13-05-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed to accept SVL™ stirrer shafts with ground glass surfaces.
  • Borosilicate glass complete with fittings
  • Glands manufactured from PTFE based polymer which is self lubricating and corrosion-resistant
  • Two types available: With SVL™ screwthread to allow connection to SVL™ flasks, With Quickfit™ ground glass cone to allow connection to Quickfit™ flasks
Specifications
Standard Taper Joint 29/32
Material Borosilicate Glass