Learn More
SVL™ Sealing Ring for Butt Joint
Product Code. 12977016
Change view
Click to view available options
Joint Size:
15
22
30
42
Unit Size:
Each
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Joint Size unitSize
12977016 42 Pack of 10
10580291 22 Pack of 10
12691591 30 Pack of 10
12681591 15 Pack of 10
12093948 15 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12977016 Supplier SVL Supplier No. 70104
Only null left
Add to basket
Add to basket
Manufactured from silicone rubber
  • Silicone rubber and sheathed with PTFE for protection against chemical attack
  • Internal diameter of ring corresponds to the bore of the SVL glass screwthread so that flow is not restricted
TRUSTED_SUSTAINABILITY
Specifica
Description Joint size: 42
For Use With (Application) Butt joints
Joint Size 42
Material Silicone Rubber, PTFE (Sheath)
Type Seal Ring
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.