Learn More
SVL™ Torion Straight Pattern Stopcocks
Manufactured from borosilicate glass
Brand: SVL 722-53
1064.00 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
- Utilizes Torion joint consisting of a Torion connector, silicone rubber sealing faced with PTFE and a PTFE key
- Vacuum capability to 10-2 Torr
- Sidearms are 70mm in length
Specifications
Stopcock | |
Borosilicate Glass | |
70 mm |
14.7 mm, Side Arm: 10 mm | |
14 mm | |
or Torion Joint Consisting of a Torion Connector, Silicone Rubber Sealing Faced with PTFE Key |