Learn More
SVL™ Torion Straight Pattern Stopcocks
Manufactured from borosilicate glass
545.00€ - 1120.00€
Specifications
Material Borosilicate Glass
View More Specs
Products 3
Product Code Brand Plug Bore Price Quantity & Availability  
Product Code Brand Plug Bore Price Quantity & Availability  
15757967
View Documents Promotion Details
SVL
722-51
5.7 mm, Side Arm: 3 mm
545.00€
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
15767967
View Documents Promotion Details
SVL
722-52
8.7 mm, Side Arm: 5 mm
685.00€
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
15577750
View Documents Promotion Details
SVL
722-53
14.7 mm, Side Arm: 10 mm
1120.00€
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Description
Description
  • Utilizes Torion joint consisting of a Torion connector, silicone rubber sealing faced with PTFE and a PTFE key
  • Vacuum capability to 10-2 Torr
  • Sidearms are 70mm in length
Specifications
Specifications
Borosilicate Glass