missing translation for 'onlineSavingsMsg'
Learn More

SVL™ Torion Straight Pattern Stopcocks

Manufactured from borosilicate glass

Brand:  SVL 722-52

650.75 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.



Additional Details : Weight : 0.27000kg

 View more versions of this product

Product Code. 15767967

  • 685.00€ / Pack of 5
Estimated Shipment:
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Utilizes Torion joint consisting of a Torion connector, silicone rubber sealing faced with PTFE and a PTFE key
  • Vacuum capability to 10-2 Torr
  • Sidearms are 70mm in length
Specifications

Specifications

Straight Pattern
8.7 mm, Side Arm: 5 mm
Borosilicate Glass
8 mm
Product Suggestions

Product Suggestions

Videos
FDS
Documents

Documents

Certificats
Promotions

Promotions