Learn More
Invitrogen™ Syntaxin 6 Recombinant Rabbit Monoclonal Antibody (1F7I2)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557995
Description
Immunogen sequence: MSMEDPFFVV KGEVQKAVNT AQGLFQRWTE LLQDPSTATR EEIDWTTNEL RNNLRSIEWD LEDLDETISI VEANPRKFNL DATELSIRKA FITSTRQVVR DMKDQMSTSS VQALAERKNR QALLGDSGSQ NWSTGTTDKY GRLDRELQRA NSHFIEEQQA QQQLIVEQQD EQLELVSGSI GVLKNMSQRI GGELEEQAVM LEDFSHELES TQSRLDNVMK KLAKVSHMTS DRRQ.
Syntaxin 6 is a member of the syntaxin superfamily. Syntaxins are nervous system-specific proteins implicated in the docking of synaptic vesicles with the presynaptic plasma membrane. Syntaxins possess a single C-terminal transmembrane domain, a SNARE domain, and an N-terminal regulatory domain. Syntaxins bind synaptotagmin in a calcium-dependent fashion and interact with voltage dependent calcium and potassium channels via the C-terminal H3 domain. Syntaxin is a key molecule in ion channel regulation and synaptic exocytosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| Syntaxin 6 | |
| Recombinant Monoclonal | |
| 1.04 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O43752, Q63635, Q9JKK1 | |
| STX6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-234 of human Syntaxin 6 (NP_001273139.1). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1F7I2 | |
| Unconjugated | |
| STX6 | |
| 2310039E05Rik; 2410005I16Rik; AA437559; AI415714; AI845861; HGNC:11441; STX6; Syn6; syntaxin 6; syntaxin-6 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10228, 58244, 60562 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.