Learn More
TFA Dostmann™ Analogue Thermo-Hygrometer with Brass Ring
Product Code. 10057641
Change view
Click to view available options
Diameter (Metric) Outer:
102 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer unitSize
10057641 102 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10057641 Supplier TFA Dostmann™ Supplier No. 45.2007
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
This thermo-hygrometer in the form of a classic pocket watch with brass bezel is an ideal measuring instrument for monitoring the room climate by keeping an eye on temperature and humidity and regulating them with targeted heating and ventilation when the values are outside the marked comfort levels.
  • Manufactured entirely in Germany
  • For a healthy indoor climate
  • Synthetic hair precision hygrometer
  • Housing with brass bezel
  • With marked comfort levels
  • No batteries required
Specifications
Material Plastic, Brass
Mounting Wall
Length (Metric) 102 mm
Width (Metric) 35 mm
Height (Metric) 113 mm
Diameter (Metric) Outer 102 mm
Weight (Metric) 65 g
Includes Instruction manual