missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYPL1 Antibody (S02-9K7), Novus Biologicals™
Rabbit Monoclonal Antibody
444.00€ - 680.00€
Specifications
| Antigen | SYPL1 |
|---|---|
| Clone | S02-9K7 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 1:20-1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18695054
|
Novus Biologicals
NBP3-19976-100ul |
100 μg |
680.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618824
|
Novus Biologicals
NBP3-19976-50ul |
50 μg |
444.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SYPL1 Monoclonal antibody specifically detects SYPL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SYPL1 | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 6856 | |
| IgG | |
| Affinity purified |
| S02-9K7 | |
| Western Blot, Immunohistochemistry 1:20-1:100, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| H-SP1, Pantophysin, synaptophysin-like 1, synaptophysin-like protein, synaptophysin-like protein 1, SYPL | |
| Recombinant protein of human SYPL1 (Uniprot # Q16563) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title