missing translation for 'onlineSavingsMsg'
Learn More

Syringaldehyde, TRC

Product Code. 30411226
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411226 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411226 Supplier Toronto Research Chemicals Supplier No. TRCS92004510G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Syringaldehyde
CAS 134-96-3
Molecular Formula C9 H10 O4
Quantity 10 g
Synonym Benzaldehyde','4-hydroxy-3,5-dimethoxy-, 4-Hydroxy-3,5-dimethoxybenzaldehyde, 2,6-Dimethoxy-4-formylphenol, 3,5-Dimethoxy-4-hydroxybenzaldehyde, 4-Formyl-2,6-dimethoxyphenol, 4-Formylsyringol, Cedar aldehyde, Gallaldehyde 3,5-dimethyl ether, NSC 41153, SM 707, Syringaldehyde, Syringic aldehyde, VND 3207
InChI Formula InChI=1S/C9H10O4/c1-12-7-3-6(5-10)4-8(13-2)9(7)11/h3-5,11H,1-2H3
SMILES COc1cc(C=O)cc(OC)c1O
IUPAC Name 4-hydroxy-3,5-dimethoxybenzaldehyde
Molecular Weight (g/mol) 182.17
Formula Weight 182.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.