Learn More
Hamilton™ Custom Cemented Needle Syringes
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
25 mL
Product Type:
Gastight
Syringe Needle
Unit Size:
Each
Description
- Ideal for dispensing volumes from 1 up to 500 μL
- A variety of volumes, lengths, and gauges available
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Product Type | Syringe Needle |
| Model | 1750 |
| Quantity | 1 Ea. |
| Volume (Metric) | 500 μL |