Learn More
CELLPATH SYSTEM II+ 2 WELL CASSETTE
Product Code. 30226256
Change view
Click to view available options
Material:
Hoppered
Loose
Threaded
Unit Size:
Pack of 1000
Pack of 1500
Pack of 2000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
30226256 Loose Pack of 1000
30226257 Hoppered Pack of 1500
30226258 Threaded Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30226256 Supplier CELLPATH Supplier No. EAM170203A
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
The System II+ 2 Well Cassettes incorporate two wells, which allows multiple biopsies to process simultaneously and significantly reduce workload. This method negates using tissue wrap or biopsy bags and makes it easier to handle biopsies, which also reduces the risk of potential core fragmentation.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Specifications
Color White
Material Loose
Product Type Cassette