Learn More
CELLPATH SYSTEM III SLOTTED CASSETTE
Product Code. 30226235
Change view
Click to view available options
Material:
Slotted
Threaded]
Unit Size:
Pack of 2000
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
30226235 Slotted Pack of 2000
30226234 Threaded] Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30226235 Supplier CELLPATH Supplier No. EAI050272B
  • ONLINE EXCLUSIVE PRICE
    360.57€
    245.96€ / Pack of 2000
    Save 114.61€
    31% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The System III Slotted Cassette comes with an attached lid with an easy to use 'break-off' hinge. With a slotted design, the cassette has a large open surface area to increase reagent flow and improve processing.
  • The cassette has a precisely engineered writing surface which optimises the print quality on inkjet and thermal printers, including the Primera, Leica and Sakura Printers.
  • Key Benefits:
  • Compatible with Primera, Leica and Sakura Printers
  • Optimised writing surface
  • Increased reagent flow
  • Integral fixed frangible lid
  • High rigidity and stability
Specifications
Color White
Material Slotted
Product Type Cassette