Learn More
SYT4 Monoclonal Antibody (OTI3E10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA505846
Encompass_Preferred
 View more versions of this product
Product Code. 17544862
  • 540.87€ / 100µL
Description
May be involved in Ca(2+)-dependent exocytosis of secretory vesicles through Ca(2+) and phospholipid binding to the C2 domain or may serve as Ca(2+) sensors in the process of vesicular trafficking and exocytosis.
Specifications
Specifications
SYT4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9H2B2
SYT4
Full length human recombit protein of human SYT4 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Western Blot
OTI3E10
Unconjugated
SYT4
HsT1192
Mouse
Affinity Chromatography
RUO
6860
-20° C, Avoid Freeze/Thaw Cycles