Learn More
SYT4 Monoclonal Antibody (OTI4F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505863
Description
May be involved in Ca(2+)-dependent exocytosis of secretory vesicles through Ca(2+) and phospholipid binding to the C2 domain or may serve as Ca(2+) sensors in the process of vesicular trafficking and exocytosis.Specifications
| SYT4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H2B2 | |
| SYT4 | |
| Full length human recombit protein of human SYT4 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI4F2 | |
| Unconjugated | |
| SYT4 | |
| HsT1192 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6860 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.