missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ T 0070907
GREENER_CHOICE

Product Code. 18736356
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18736356 5 mg 5mg
18709516 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18736356 Supplier Tocris Bioscience™ Supplier No. 2301/5
  • ONLINE EXCLUSIVE PRICE
    146.00€
    137.55€ / 5mg
    Save 8.45€
    5% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Highly potent and selective covalent PPARγ antagonist

T 0070907 is a potent and selective irreversible PPARγ antagonist (IC50 = 1 nM). Displays > 800-fold selectivity for PPARγ over PPARα and PPARδ. Blocks transcriptional activity of PPARγ in vitro and inhibits Rosiglitazone -induced adipogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloro-5-nitro-N-4-pyridinylbenzamide
CAS 313516-66-4
Assay Percent Range >99%
Quantity 5 mg
Molecular Formula C12H8N3O3Cl
Formula Weight Observed MW: 277.67
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.