missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T-bet/TBX21 Antibody (525803) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5385JF585
This item is not returnable.
View return policy
Description
T-bet/TBX21 Monoclonal antibody specifically detects T-bet/TBX21 in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),Immunocytochemistry
Specifications
| T-bet/TBX21 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM | |
| E. coli-derived recombinant human T-bet, Glu326-Asn535, Accession # Q9UL17 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| 525803 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Immunology | |
| 30009 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction