Learn More
Copan Italia™ T-Shaped Plastic Spreaders
Description
- Used for performing bacterial CFU plate counts on water, milk, urine, and other liquid samples.
- Smooth, rounded surfaces prevent cutting or gouging of agar during inoculation of culture plates.
- T-shape design facilitates even pressure application on the agar surface.
Specifications
Specifications
| Disposable | Single-use |
| Length (Metric) | 171 mm |
| Packaging | 5/Peel pack, 1000/Cs |
| Quantity | 1000/Pk. |
| Shape | T |
| Material | Plastic |
| Product Type | Spreader |
| Sterility | Sterile |
| For Use With (Application) | Used for performing bacterial CFU plate counts on water, milk, urine and other liquid samples. |