missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TADA3L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10839-1-AP-150UL
This item is not returnable.
View return policy
Description
Many DNA-binding transcriptional activator proteins enhance the initiation rate of RNA polymerase II-mediated gene transcription by interacting functionally with the general transcription machinery bound at the basal promoter. Adaptor proteins are usually required for this activation, possibly to acetylate and destabilize nucleosomes, thereby relieving chromatin constraints at the promoter. The protein encoded by this gene is a transcriptional activator adaptor and has been found to be part of the PCAF histone acetylase complex. In addition, it associates with the tumor suppressor protein p53 and is required for full activity of p53 and p53-mediated apoptosis. At least four alternatively spliced variants have been found for this gene, but the full-length nature of some variants has not been determined.Specifications
| TADA3L | |
| Polyclonal | |
| Unconjugated | |
| TADA3 | |
| ADA3, ADA3 homolog, ADA3 like protein, hADA3, STAF54, TADA3, TADA3L, Transcriptional adapter 3, Transcriptional adapter 3 like | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 101206, 10474, 362414 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75528, Q4V8F5, Q8R0L9 | |
| Tada3 | |
| TADA3L Fusion Protein Ag1276 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction