Learn More
Tanshinone IIA, TRC
Product Code. 30425979
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425979 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30425979 Supplier Toronto Research Chemicals Supplier No. TRCT007000100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tanshinone IIA
CAS 568-72-9
Molecular Formula C19 H18 O3
Quantity 100 mg
Synonym 6,7,8,9-Tetrahydro-1,6,6-trimethyl-phenanthro[1,2-b]furan-10,11-dione, Tanshinone IIA (7CI), 1,6,6-Trimethyl-6,7,8,9-tetrahydrophenanthro[1,2-b]furan-10,11-dione, Dan Shen ketone, NSC 686518, NSC 686519, Salviol IIA, Tanshinon II, Tanshinone B, Tanshinone II, Tashinone IIA
InChI Formula InChI=1S/C19H18O3/c1-10-9-22-18-12-6-7-13-11(5-4-8-19(13,2)3)15(12)17(21)16(20)14(10)18/h6-7,9H,4-5,8H2,1-3H3
SMILES Cc1coc2c3ccc4c(CCCC4(C)C)c3C(=O)C(=O)c12
IUPAC Name 1,6,6-trimethyl-8,9-dihydro-7H-naphtho[1,2-g][1]benzofuran-10,11-dione
Molecular Weight (g/mol) 294.34
Formula Weight 294.1256