Learn More
Tantalum(V) tetraethoxide 2,4-pentanedionate, 99.99%, (metals basis)
Description
Tantalum(V) tetraethoxide 2,4-pentanedionate is used in pharmaceutical, biochemical and chemical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 45°C |
| Density | 1.5 g/mL |
| Boiling Point | 95°C (0.5 mmHg) |
| Flash Point | >65°C (149°F) |
| Quantity | 50 g |
| Sensitivity | Moisture sensitive |
| Solubility Information | Reacts with water. |
| IUPAC Name | tantalum(5+) (2Z)-4-oxopent-2-en-2-olate tetraethanolate |
| Formula Weight | 460.3 |
| Percent Purity | 99.99% |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.