Learn More
TAPBPL Mouse, anti-Human, Clone: 1D12C2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68577-1-IG-150UL
 View more versions of this product
Product Code. 18007415
  • 410.00€ / 150µL
Description
The TAPBPL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TAPBPL. This antibody recognizes human antigen. The TAPBPL antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TAPBPL
1D12C2
FLJ10143, TAP binding protein like, Tapasin like, TAPASIN R, Tapasin related protein, TAPBP R, TAPBPL, TAPBPR
150 μL
55080
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human
150 kDa