Learn More
TAPBPL Mouse, anti-Human, Clone: 1D12C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68577-1-IG-150UL
Description
The TAPBPL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TAPBPL. This antibody recognizes human antigen. The TAPBPL antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TAPBPL | |
| 1D12C2 | |
| FLJ10143, TAP binding protein like, Tapasin like, TAPASIN R, Tapasin related protein, TAPBP R, TAPBPL, TAPBPR | |
| 150 μL | |
| 55080 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |