missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Taps, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Taps |
| CAS | 29915-38-6 |
| Molecular Formula | C7 H17 N O6 S |
| Quantity | 5 g |
| Synonym | N-[Tris(hydroxymethyl)methyl]-3-aminopropanesulfonic acid','[(2-Hydroxy-1,1-bis(hydroxymethyl)ethyl)amino]-1-propanesulfonic acid |
| InChI Formula | InChI=1S/C7H17NO6S/c9-4-7(5-10,6-11)8-2-1-3-15(12,13)14/h8-11H,1-6H2,(H,12,13,14) |
| SMILES | OCC(CO)(CO)NCCCS(=O)(=O)O |
| IUPAC Name | 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid |
| Molecular Weight (g/mol) | 243.278 |
| Formula Weight | 243.078 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction