missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Taq DNA Polymerase Monoclonal Antibody (3C3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA542391
Additional Details : Weight : 0.01000kg
Description
Used in the PCR method because of its high thermostability. Has a relatively high error rate because it lacks exonuclease proofreading functionality [Uniprot].Specifications
Taq DNA Polymerase | |
Monoclonal | |
2 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
Taq DNA Polymerase | |
100 μL | |
Primary | |
Antibody | |
IgG1 |
Functional Assay | |
3C3 | |
Unconjugated | |
Mouse | |
Protein G | |
RUO | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Chemical |