Learn More
MP Biomedicals™ Taq Pol Bulk
Product Code. 11467010 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 x 250U
1000U
250U
3 x 250U
5 x 1000U
Unit Size:
1000 units
250 units
750 units
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467010 250U 250 units
11497010 3 x 250U 750 units
11477010 1000U 1000 units
11417020 10 x 250U 250 units
11487010 5 x 1000U 1000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11467010 Supplier MP Biomedicals™ Supplier No. 11EPTQA025
  • ONLINE EXCLUSIVE PRICE
    186.00€
    176.40€ / 250 units
    Save 9.60€
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket
For life science applications
  • Ready for CE IVD labeling of clinical applications
TRUSTED_SUSTAINABILITY
Specifications
Concentration 5 U/μL
Description Taq Pol Bulk
Quantity 250U
Includes Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2
For Resaerch Use Only