missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TaqMan™ Cells-to-CT™ HepatoExpress™ Kit

Product Code. 31722965
Change view
Click to view available options
No. of Reactions:
100 Reactions
2500 Reactions (Lysis only)
40 Reactions
400 Reactions
Unit Size:
1 set
100 reactions
40 reactions
400 reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
31722965 40 Reactions 40 reactions
31722703 100 Reactions 100 reactions
31722752 400 Reactions 400 reactions
31722757 2500 Reactions (Lysis only) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31722965 Supplier Invitrogen™ Supplier No. A65788

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The TaqMan Cells-to-CT HepatoExpress Kit is a complete cell lysate system designed for gene expression analysis directly from hepatocytes without the need of RNA purification.

The TaqMan Cells-to-CT HepatoExpress Kit offers a complete cell lysis and RT-qPCR workflow solution designed for high-throughput gene expression analysis directly from cultured primary hepatocytes without the need for RNA purification. This innovative kit includes HepatoExpress Lysis Solution, HepatoExpress LysisGuard reagent, ezDNase enzyme, SuperScript IV VILO Master Mix, and the TaqMan Fast Advanced Master Mix. The newly developed HepatoExpress Lysis Solution and LysisGuard reagents enable superior lysis performance and sample stability, while the best-in-class RT-qPCR reagents deliver high sensitivity for a seamless end-to-end workflow in approximately 75 minutes.

Features of the TaqMan Cells-to-CT HepatoExpress Kit include:

  • Effortless workflow—simple process that supports high-throughput processing and automation
  • No tedious RNA purification—no columns, beads, heating, or centrifugation
  • Enhanced stability and lysis—efficient primary hepatocyte lysis, while preserving RNA
  • Robust performance—achieve results equivalent to those from purified RNA
  • Maximum flexibility—accommodates extracellular matrix overlays and multiplexing of targets
  • Sustainable solution—REACH-compliant reagents and minimized plastic waste

Optimized chemistry without sacrificing sensitivity

Gene expression analysis from primary hepatocytes is now simplified with our TaqMan Cells-to-CT HepatoExpress Kit. With a strong lysis that penetrates extracellular matrix overlays without degrading RNA, this product enables a rapid workflow without compromising sensitivity. The inclusion of ezDNase enables integrated removal of genomic DNA during cell lysis. After a 10-minute incubation at room temperature, the lysate is ready for reverse transcription using SuperScript IV VILO Master Mix. Once the RT reaction is complete, qPCR analysis is conducted using the TaqMan Fast Advanced qPCR Master Mix and a TaqMan Gene Expression Assay.

Streamlined workflow with proven performance

RNA prepared using the TaqMan Cells-to-CT HepatoExpress technology substantially expedites and streamlines gene expression analysis of primary hepatocytes. Cell lysates exhibit sensitivity and specificity equivalent to RNA extracted using traditional spin column isolation methods.

The lysis procedure simultaneously prepares cell lysates for RT-qPCR and removes genomic DNA in only ten minutes. The lysis step is easily automated with robotic platforms for high-throughput processing of 96-well plates, as it occurs entirely at room temperature. The high-efficiency SuperScript IV VILO RT Master Mix linearly transcribes RNA across a broad input range, even with suboptimal template purity. Additionally, the TaqMan Cells-to-CT HepatoExpress Kit enables multiplexed detection of targets and is compatible with over 700,000 TaqMan Gene Expression Assays.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage HepatoExpress Lysis Solution (Room Temperature), HepatoExpress LysisGuard (-20°C), Express ezDNase (-20°C), SuperScript IV VILO Master Mix (-20°C), SuperScript IV VILO No RT Control (-20°C), TaqMan Fast Advanced Master Mix (4°C). For long-term storage, store HepatoExpress Lysis Buffer at 4°C.
Detection Method TaqMan
Form Liquid
Format Kit
GC-Rich PCR Performance High
PCR Method 2-step RT-qPCR
Reaction Speed 10 min.
High-throughput Compatibility Yes
Label or Dye ROX
Product Type Direct Cell Lysis to RT-qPCR Kit
Quantity 40 Reactions
Shipping Condition Frozen, Dry Ice
Target cDNA
For Use With (Application) Gene Expression
Isolation Technology Direct Cell Lysis
No. of Reactions 40 Reactions
Purification Target RNA
Sample Type Hepatocytes, HepaRG, HepG2
Starting Material Up to 100,000 Cultured Cells
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.