missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ hPSC Scorecard™ Panel, Fast 96-well

Product Code. 15343826 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
2 x 96-well plates
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15343826 2 x 96-well plates Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15343826 Supplier Applied Biosystems™ Supplier No. A15876
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

TaqMan™ hPSC Scorecard™ Panel, Fast 96-well

The Applied Biosystems™ TaqMan™ hPSC Scorecard™ Panel 2 x 96w FAST enables verification of pluripotency and determination of lineage bias for both ES and iPS cell lines. Each of the two 96-well plates contains 94 predefined TaqMan™ Gene Expression assays (including endogenous controls) dried-down in the wells. Master mix is not included with this panel. For a kit that combines the panel with master mix, see TaqMan™ hPSC Scorecard™ Kit 2 x 96w FAST. This product is also available in a 384-well format.

  • Assess an entire gene signature in one simple experiment
  • Compare results to a reference standard with the accompanying hPSC Scorecard™ Analysis Software
  • Identify lines with trilineage differentiation potential
  • Evaluate inductive differentiation specified to one germ layer
  • Access validated content for increased confidence in your results

    Assess an Entire Gene Signature in One Experiment
    The TaqMan™ hPSC Scorecard™ Panel helps save time by offering pre-plated assays in a stable and convenient dried-down format. Simply add TaqMan™ Master Mix to your cDNA of interest and transfer to the 96-well plate with a multichannel pipette.

    Compare Results to a Reference Standard
    The TaqMan™ hPSC Scorecard™ Panel includes access to proprietary cloud-based analysis software that allows you to view and export graphs and tables of results, as well as generate reports. The software is compatible with seven Applied Biosystems™ qRT-PCR systems and is available at no additional charge.

    Identify Lines with Trilineage Differentiation Potential
    Include randomly differentiated embryoid body (EB) samples (differentiated for at least 7 days using standard EB methods) in your analysis and determine whether your lines are biased toward one or more of the three germ layers (endoderm, mesoderm, ectoderm).

    Evaluate Inductive Differentiation Specified to One Germ Layer
    The panel includes over twenty markers for each of the three germ layers that have been shown to respond as expected to directed differentiation in monolayer culture. Quickly evaluate time course data, culture conditions and media formulations for your germ layer of interest with this one panel.

    Access Validated Content
    The panel contents are based on published work (Bock et al., Cell 144, 439–452, 2011) and were validated against multiple human ES and iPS lines.

    What You Get
    The TaqMan™ hPSC Scorecard™ Panel 2 x 96w FAST includes:

  • Two 96-well plates of 94 TaqMan™ Gene Expression assays
  • Two optical plate covers
  • TRUSTED_SUSTAINABILITY

    Specifications

    Type Panel
    Format 96-array Plate
    Species Human
    Content And Storage Contents: Two 96-well plates each containing the 96-assay panel and two optical covers

    Store at room temperature.
    Detection Method Primer-Probe Detection
    Form Dried Down
    For Use With (Equipment) QuantStudio™ 12k Flex, StepOne™, Fast Mode, ViiA™ 7 System
    Includes 2 x 96-well plate
    Product Line TaqMan
    Quantity 2 x 96-well plates
    Reaction Speed Fast
    Research Category Stem Cell Research
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.