missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TARC Monoclonal Antibody (110946)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561542
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
TARC ligand (CCL17) belongs to the CC chemokine family. It is a small cytokine and the gene is located on q arm of chromosome 16, in humans, along with other chemokines. TARC specifically binds to T-cells and interacts with chemokine receptors CCR4 and CCR8. It plays an important role in T cell development in thymus as well as in trafficking and activation of mature T cells.
Specifications
| TARC | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| A-152E5.3; ABCD-2; CC chemokine TARC; C-C motif chemokine; C-C motif chemokine 17; C-C motif chemokine ligand 17; CCL17; chemokine (C-C motif) ligand 17; SCYA17; Scya17l; small inducible cytokine subfamily A (Cys-Cys), member 17; small inducible cytokine subfamily A17; small-inducible cytokine A17; T cell-directed CC chemokine; TARC; Thymus and activation-regulated chemokine | |
| Rat | |
| Protein A/G | |
| RUO | |
| 20295 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 110946 | |
| Unconjugated | |
| CCL17 | |
| CCL17 | |
| E. coli-derived recombinant mouse CCL17/TARC, Ala24-Pro93. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction