missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS1R1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA132846
Additional Details : Weight : 0.01000kg
Description
PA1-32846 detects GPCR TAS1R1 from human samples.
Putative taste receptor. TAS1R1/TAS1R3 responds to the umami taste stimulus (the taste of monosodium glutamate) and also to most of the 20 standard L-amino acids, but not to their D-enantiomers or other compounds. Sequence differences within and between species can significantly influence the selectivity and specificity of taste responses. Expressed strongly only in fungiform papillae.Specifications
TAS1R1 | |
Polyclonal | |
Unconjugated | |
TAS1R1 | |
candidate taste receptor T1R1; G protein-coupled receptor 70; GM148; GPCRB3; GPCR-B3; GPR70; G-protein coupled receptor 70; seven transmembrane helix receptor; T1r1; TAS1R1; taste 1 receptor member 1; taste receptor type 1 member 1; taste receptor, type 1, member 1; TR1; TS1R1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80835 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q7RTX1 | |
TAS1R1 | |
Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human TAS1R1 | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |